Learn More
TARGETMOL CHEMICALS INC BMS CCR2 22 5MG
SDP
Catalog No. 502581280
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMS CCR2 22 is a potent and selective antagonist of CCR2 with calcium flux IC50 of 18 nM chemotaxis IC50 of 1 nM and binding IC50 of 5.1 nM. purity: 98%
Catalog No. 50-258-1280 Supplier TARGETMOL CHEMICALS INC Supplier No. T146885MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMS CCR2 22 is a potent and selective antagonist of CCR2 with calcium flux IC50 of 18 nM chemotaxis IC50 of 1 nM and binding IC50 of 5.1 nM. purity: 98%