Learn More
Apexbio Technology LLC BMS345541 hydrochloride, 10mg. Cas: 547757-23-3 MFCD: MFCD07784499. IKK inhibitor,highly selective
SDP
Supplier:  Apexbio Technology LLC A324810
Encompass_Preferred
BMS-345541 is a highly selective inhibitor of IKK-1 and IKK-2 with IC50 values of 4M and 0.3M, respectively [1].BMS-345541 is a highly selective inhibitor of IKK that inhibits NF-B-dependent transcription of pro-inflammatory cytokines both in vitro and in vivo. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4792