Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BMT-090605 hydrochloride | 2231664-45-0 | 98.8% | 400.90 g/mol | C21H25ClN4O2 | 1 ML
SDP

Catalog No. 5000211668
Encompass_Preferred

This hydrochloride salt is a small-molecule kinase inhibitor targeting AAK1 and GAK, supplied for research use in both solid and solution formats suitable for in vitro and cell-based assays.

  • High purity (98.8%).
  • Available as 10 mM, 1 mL solution in DMSO and as solids (5-100 mg).
  • Good solubility in DMSO: ~31.25 mg/mL.
  • Molecular weight 400.90 g/mol; molecular formula C21H25ClN4O2.
  • Recommended storage: powder at -20°C; stock solutions at -80°C.

Catalog No. 50-002-11668 Supplier Medchemexpress LLC Supplier No. HY101290A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This hydrochloride salt is a small-molecule kinase inhibitor targeting AAK1 and GAK, supplied for research use in both solid and solution formats suitable for in vitro and cell-based assays.

  • High purity (98.8%).
  • Available as 10 mM, 1 mL solution in DMSO and as solids (5-100 mg).
  • Good solubility in DMSO: ~31.25 mg/mL.
  • Molecular weight 400.90 g/mol; molecular formula C21H25ClN4O2.
  • Recommended storage: powder at -20°C; stock solutions at -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.