Learn More
Medchemexpress LLC BMT-090605 (hydrochloride) | 2231664-45-0 | 98.8% | 400.90 g/mol | C21H25ClN4O2 | 10 MG
SDP
Catalog No. 5000211669
Encompass_Preferred
BMT-090605 hydrochloride is a research-grade small-molecule hydrochloride salt that functions as a potent inhibitor of adaptor protein-2 associated kinase 1 (AAK1), with reported activity against BIKE and GAK. It is supplied as a solid for use in in vitro kinase assays and preclinical pharmacology studies.
  • Potent AAK1 inhibitor with low-nanomolar activity.
  • Also inhibits BIKE and GAK, supporting broad kinase profiling.
  • High purity (≈98.8%) suitable for research applications.
  • Supplied as a solid light yellow powder for convenient handling.
  • Stable when stored at -20°C as a powder for long-term storage.
  • Packaged in small milligram quantities for laboratory use.
Catalog No. 50-002-11669 Supplier Medchemexpress LLC Supplier No. HY101290A10MG
BMT-090605 hydrochloride is a research-grade small-molecule hydrochloride salt that functions as a potent inhibitor of adaptor protein-2 associated kinase 1 (AAK1), with reported activity against BIKE and GAK. It is supplied as a solid for use in in vitro kinase assays and preclinical pharmacology studies.
  • Potent AAK1 inhibitor with low-nanomolar activity.
  • Also inhibits BIKE and GAK, supporting broad kinase profiling.
  • High purity (≈98.8%) suitable for research applications.
  • Supplied as a solid light yellow powder for convenient handling.
  • Stable when stored at -20°C as a powder for long-term storage.
  • Packaged in small milligram quantities for laboratory use.