Learn More
Medchemexpress LLC Bmt-090605 (hydrochloride) | 2231664-45-0 | 98.8% | 400.90 g/mol | C21H25ClN4O2 | 100 MG
SDP
Catalog No. 5000211286
Encompass_Preferred
BMT-090605 hydrochloride is the hydrochloride salt of a potent small-molecule AAK1 inhibitor supplied for research use. It is highly characterized, with confirmed identity and stability data supporting preclinical applications such as antinociceptive and neuropathic pain studies.
  • Potent AAK1 inhibitor (IC50 0.6 nM).
  • Also inhibits BIKE and GAK (IC50 45 nM and 60 nM).
  • High purity (98.8%) with verified analytical data.
  • Available as solid and solution formulations in multiple sizes, including 100 mg.
  • Recommended storage: powder at -20°C for long-term; in solvent at -80°C for best stability.
Catalog No. 50-002-11286 Supplier Medchemexpress LLC Supplier No. HY101290A100MG
BMT-090605 hydrochloride is the hydrochloride salt of a potent small-molecule AAK1 inhibitor supplied for research use. It is highly characterized, with confirmed identity and stability data supporting preclinical applications such as antinociceptive and neuropathic pain studies.
  • Potent AAK1 inhibitor (IC50 0.6 nM).
  • Also inhibits BIKE and GAK (IC50 45 nM and 60 nM).
  • High purity (98.8%) with verified analytical data.
  • Available as solid and solution formulations in multiple sizes, including 100 mg.
  • Recommended storage: powder at -20°C for long-term; in solvent at -80°C for best stability.