Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BMT-090605 hydrochloride | 2231664-45-0 | 98.8% | 400.90 g/mol | C21H25ClN4O2 | 25 MG
SDP

Catalog No. 5000212652
Encompass_Preferred

BMT-090605 hydrochloride is the hydrochloride salt of a potent kinase inhibitor selective for adaptor protein-2 associated kinase 1 (AAK1). It is supplied for preclinical research use, including studies of neuropathic pain, and shows activity versus related kinases in addition to AAK1.

  • Potent AAK1 inhibitor (IC50 0.6 nM).
  • Activity against BIKE and GAK (IC50 45 nM and 60 nM).
  • High purity (98.8%).
  • Molecular weight 400.90 g/mol; formula C21H25ClN4O2.
  • Available as a solid in multiple sizes, including 25 mg.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.

Catalog No. 50-002-12652 Supplier Medchemexpress LLC Supplier No. HY101290A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMT-090605 hydrochloride is the hydrochloride salt of a potent kinase inhibitor selective for adaptor protein-2 associated kinase 1 (AAK1). It is supplied for preclinical research use, including studies of neuropathic pain, and shows activity versus related kinases in addition to AAK1.

  • Potent AAK1 inhibitor (IC50 0.6 nM).
  • Activity against BIKE and GAK (IC50 45 nM and 60 nM).
  • High purity (98.8%).
  • Molecular weight 400.90 g/mol; formula C21H25ClN4O2.
  • Available as a solid in multiple sizes, including 25 mg.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.