Learn More
Medchemexpress LLC Bmt-090605 (hydrochloride) | 2231664-45-0 | 98.8% | 400.90 g·mol⁻¹ | C21H25ClN4O2 | 5 MG
SDP
Catalog No. 5000211354
Encompass_Preferred
BMT-090605 hydrochloride is a potent, selective small-molecule inhibitor of adaptor protein-2 associated kinase 1 (AAK1) with additional activity against BIKE and GAK. Supplied as the hydrochloride salt for research use, it is provided as a solid with specified purity and storage recommendations, making it suitable for kinase profiling and target validation studies.
  • Potent AAK1 inhibition with low-nanomolar IC50 (0.6 nM).
  • Selective activity against related kinases BIKE and GAK.
  • High purity (98.8%) suitable for biochemical assays.
  • Molecular weight 400.90 g·mol⁻¹.
  • Available in small pre-weighed sizes for research, including 5 mg.
  • Stable as a powder when stored at recommended temperatures.
Catalog No. 50-002-11354 Supplier Medchemexpress LLC Supplier No. HY101290A5MG
BMT-090605 hydrochloride is a potent, selective small-molecule inhibitor of adaptor protein-2 associated kinase 1 (AAK1) with additional activity against BIKE and GAK. Supplied as the hydrochloride salt for research use, it is provided as a solid with specified purity and storage recommendations, making it suitable for kinase profiling and target validation studies.
  • Potent AAK1 inhibition with low-nanomolar IC50 (0.6 nM).
  • Selective activity against related kinases BIKE and GAK.
  • High purity (98.8%) suitable for biochemical assays.
  • Molecular weight 400.90 g·mol⁻¹.
  • Available in small pre-weighed sizes for research, including 5 mg.
  • Stable as a powder when stored at recommended temperatures.