Learn More
Medchemexpress LLC Bmt-090605 hydrochloride | 2231664-45-0 | 98.8% | 400.90 g/mol | C21H25ClN4O2 | 50 MG
SDP
Catalog No. 5000211353
Encompass_Preferred
BMT-090605 hydrochloride is the hydrochloride salt of a potent, selective adapter protein-2 associated kinase 1 (AAK1) inhibitor for biochemical and cellular research. Supplied as a light-yellow solid with high reported purity, it is intended for in vitro and early-stage preclinical studies to probe AAK1-mediated pathways and mechanism-of-action experiments.
  • High reported purity suitable for research-grade assays.
  • Supplied as a solid with consistent physical characterization.
  • Available in small research pack sizes, enabling dose-range studies.
  • Used to interrogate AAK1-related signaling in biochemical and cellular assays.
  • Stable under typical frozen and refrigerated storage conditions (refer to SDS).
Catalog No. 50-002-11353 Supplier Medchemexpress LLC Supplier No. HY101290A50MG
BMT-090605 hydrochloride is the hydrochloride salt of a potent, selective adapter protein-2 associated kinase 1 (AAK1) inhibitor for biochemical and cellular research. Supplied as a light-yellow solid with high reported purity, it is intended for in vitro and early-stage preclinical studies to probe AAK1-mediated pathways and mechanism-of-action experiments.
  • High reported purity suitable for research-grade assays.
  • Supplied as a solid with consistent physical characterization.
  • Available in small research pack sizes, enabling dose-range studies.
  • Used to interrogate AAK1-related signaling in biochemical and cellular assays.
  • Stable under typical frozen and refrigerated storage conditions (refer to SDS).