Learn More
TARGETMOL CHEMICALS INC BMT-124110 FORMATE 5MG
SDP
Catalog No. 502581708
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMT-124110 Formate is a selective AAK1 inhibitor (IC50 0.9 nM) with anti-injury sensory activity.BMT-124110 Formate inhibits the BMP-2-inducible protein kinase BIKE with an IC50 17 nM.BMT-124110 Formate inhibits the cell-cycle protein G-related BMT-124110 Formate inhibits the cell cycle protein G-associated kinase GAK with an IC5099 nM.
Catalog No. 50-258-1708 Supplier TARGETMOL CHEMICALS INC Supplier No. T10572L5MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMT-124110 Formate is a selective AAK1 inhibitor (IC50 0.9 nM) with anti-injury sensory activity.BMT-124110 Formate inhibits the BMP-2-inducible protein kinase BIKE with an IC50 17 nM.BMT-124110 Formate inhibits the cell-cycle protein G-related BMT-124110 Formate inhibits the cell cycle protein G-associated kinase GAK with an IC5099 nM.