Learn More
Medchemexpress LLC BMT-124110 (formic) | 00-00-0 | MFCD34469994 | >98.0% | 369.46 g/mol | C21H27N3O3 | 5 MG
SDP
Catalog No. 5000223508
Encompass_Preferred
Small-molecule AAK1 inhibitor for laboratory research. It is a potent, selective AAK1 inhibitor (IC50 = 0.9 nM) with reported antinociceptive activity; it also inhibits BIKE (IC50 = 17 nM) and GAK (IC50 = 99 nM). Molecular formula C21H27N3O3; molecular weight 369.46 g/mol. Store powder at -20°C; in solution store at -80°C for up to 6 months or at -20°C for up to 1 month. Purity reported >98% (HPLC).
  • Potent AAK1 inhibition (IC50 = 0.9 nM).
  • Activity against BIKE and GAK (IC50s 17 nM and 99 nM).
  • Suitable for biochemical and cellular kinase assays.
  • Stable when stored as powder at -20°C.
  • Supplied in milligram research quantities.
Catalog No. 50-002-23508 Supplier Medchemexpress LLC Supplier No. HY135871A5MG
Small-molecule AAK1 inhibitor for laboratory research. It is a potent, selective AAK1 inhibitor (IC50 = 0.9 nM) with reported antinociceptive activity; it also inhibits BIKE (IC50 = 17 nM) and GAK (IC50 = 99 nM). Molecular formula C21H27N3O3; molecular weight 369.46 g/mol. Store powder at -20°C; in solution store at -80°C for up to 6 months or at -20°C for up to 1 month. Purity reported >98% (HPLC).
  • Potent AAK1 inhibition (IC50 = 0.9 nM).
  • Activity against BIKE and GAK (IC50s 17 nM and 99 nM).
  • Suitable for biochemical and cellular kinase assays.
  • Stable when stored as powder at -20°C.
  • Supplied in milligram research quantities.