Learn More
Medchemexpress LLC HY-100728 5mg Medchemexpress, BMT-145027 CAS:2018282-44-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1007285MG
Encompass_Preferred
Medchemexpress, HY-100728 5mg BMT-145027 CAS:2018282-44-3 BMT-145027 is an mGluR5 positive allosteric modulator without inherent agonist activity, exhibits an EC 50 of 47 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1928