Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BMT-297376 | 2251031-81-7 | 99.14% | 433.49 | 1 ML
SDP

Catalog No. 5000351395
Encompass_Preferred

BMT-297376 is an optimized Linrodostat and a potent IDO1 inhibitor. This product is for research use only and is not sold to patients.

  • Purity: 99.14%
  • Molecular weight: 433.49
  • Formula: C23H29F2N3O3
  • CAS No.: 2251031-81-7
  • Appearance: Solid, white to off-white

Catalog No. 50-003-51395 Supplier Medchemexpress LLC Supplier No. HY13920510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMT-297376 is an optimized Linrodostat and a potent IDO1 inhibitor. This product is for research use only and is not sold to patients.

  • Purity: 99.14%
  • Molecular weight: 433.49
  • Formula: C23H29F2N3O3
  • CAS No.: 2251031-81-7
  • Appearance: Solid, white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.