Learn More
Apexbio Technology LLC BMX-IN-1(Synonyms: BMX Kinase Inhibitor, ETK kinase inhibitor, BMX-IN1, BMX inhibitor, ETK inhibitor), 10mM (in 1mL DMSO), CAS: 1431525-23-3.
SDP
Catalog No. 5000060287
Encompass_Preferred
BMX-IN-1 (CAS 1431525-23-3) is a highly selective irreversible inhibitor targeting BMX (bone marrow kinase on chromosome X also known as ETK) a member of the Tec tyrosine kinase family BMX is prominently expressed in arterial endothelium and myeloid hematopoietic cells functioning in ischemia-induced arterial and lymphatic vessel formation processes and contributing to tumor growth In cellular models expressing Tel-BMX fusion proteins BMX-IN-1 treatment at low doses effectively reduces proliferation highlighting its potential utility in cancer research particularly prostate cancer studies
Catalog No. 50-000-60287 Supplier Apexbio Technology LLC Supplier No. A32605.1
BMX-IN-1 (CAS 1431525-23-3) is a highly selective irreversible inhibitor targeting BMX (bone marrow kinase on chromosome X also known as ETK) a member of the Tec tyrosine kinase family BMX is prominently expressed in arterial endothelium and myeloid hematopoietic cells functioning in ischemia-induced arterial and lymphatic vessel formation processes and contributing to tumor growth In cellular models expressing Tel-BMX fusion proteins BMX-IN-1 treatment at low doses effectively reduces proliferation highlighting its potential utility in cancer research particularly prostate cancer studies