Learn More
Medchemexpress LLC BMY-14802 hydrochloride | 105565-55-7 | 98.0% | 384.85 | 500 UG
SDP
Catalog No. 5000346967
Encompass_Preferred
BMY-14802 hydrochloride (BMY-14802-1) is a selective and orally active sigma receptor antagonist with an IC50 of 112 nM. It is also a 5-HT1A and adrenergic α1 receptors agonist. BMY-14802 hydrochloride has antipsychotic effects.
  • Selective and orally active sigma receptor antagonist.
  • Acts as a 5-HT1A and adrenergic α1 receptor agonist.
  • Exhibits antipsychotic effects.
Catalog No. 50-003-46967 Supplier Medchemexpress LLC Supplier No. HY10850910MM/1ML
BMY-14802 hydrochloride (BMY-14802-1) is a selective and orally active sigma receptor antagonist with an IC50 of 112 nM. It is also a 5-HT1A and adrenergic α1 receptors agonist. BMY-14802 hydrochloride has antipsychotic effects.
  • Selective and orally active sigma receptor antagonist.
  • Acts as a 5-HT1A and adrenergic α1 receptor agonist.
  • Exhibits antipsychotic effects.