Learn More
Medchemexpress LLC BMY 14802 hydrochloride | 105565-55-7 | MFCD04039786 | 98.0% | 384.85 | C18H23ClF2N4O | 5 MG
SDP
Catalog No. 5000263720
Encompass_Preferred
BMY-14802 hydrochloride is the hydrochloride salt of a selective and orally active sigma receptor antagonist that also exhibits 5-HT1A agonist and α1-adrenergic activity. It is used in preclinical research exploring antipsychotic effects and movement disorders, and is supplied for research use only.
  • Selective sigma receptor antagonist with reported IC50 = 112 nM.
  • Also displays 5-HT1A agonist activity and α1-adrenergic effects.
  • Supplied for research use only; not for human or veterinary use.
  • High purity by HPLC (≈98.0%).
  • Available as solid and as a 10 mM solution in DMSO.
  • Molecular weight 384.85; formula C18H23ClF2N4O.
Catalog No. 50-002-63720 Supplier Medchemexpress LLC Supplier No. HY1085095MG
BMY-14802 hydrochloride is the hydrochloride salt of a selective and orally active sigma receptor antagonist that also exhibits 5-HT1A agonist and α1-adrenergic activity. It is used in preclinical research exploring antipsychotic effects and movement disorders, and is supplied for research use only.
  • Selective sigma receptor antagonist with reported IC50 = 112 nM.
  • Also displays 5-HT1A agonist activity and α1-adrenergic effects.
  • Supplied for research use only; not for human or veterinary use.
  • High purity by HPLC (≈98.0%).
  • Available as solid and as a 10 mM solution in DMSO.
  • Molecular weight 384.85; formula C18H23ClF2N4O.