Learn More
TARGETMOL CHEMICALS INC BMY-14802 HYDROCHLORIDE 5MG
SDP
Catalog No. 502580940
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. BMY-14802 hydrochloride (BMS 181100 hydrochloride) is a selective antagonist of (sigma) receptor (IC50 = 112 nM) with antipsychotic effects. BMY-14802 hydrochloride is an agonist of the (alpha)1-adrenergic receptor and 5-HT1A. purity: 98%
Catalog No. 50-258-0940 Supplier TARGETMOL CHEMICALS INC Supplier No. T226125MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. BMY-14802 hydrochloride (BMS 181100 hydrochloride) is a selective antagonist of (sigma) receptor (IC50 = 112 nM) with antipsychotic effects. BMY-14802 hydrochloride is an agonist of the (alpha)1-adrenergic receptor and 5-HT1A. purity: 98%