Learn More
Cayman Chemical BMY 14802 (hydrochloride), Purity: ≥98%, CAS: 105565-55-7, 50 mg
SDP
Catalog No. 502943501
Encompass_Preferred
An atypical antipsychotic; selectively binds to σ receptors over α1-ARs and dopamine D2 receptors (IC50s = 112, 520, and 6,340 nM, respectively); inhibits the conditioned avoidance response and apomorphine-induced stereotypy in rats (ED50s = 26.4 and 33 mg/kg, respectively)
Catalog No. 50-294-3501 Supplier Cayman Chemical Supplier No. 4226450 MG
An atypical antipsychotic; selectively binds to σ receptors over α1-ARs and dopamine D2 receptors (IC50s = 112, 520, and 6,340 nM, respectively); inhibits the conditioned avoidance response and apomorphine-induced stereotypy in rats (ED50s = 26.4 and 33 mg/kg, respectively)