Learn More
Medchemexpress LLC Bmy 28674-d8 | 1189644-16-3 | 96.1% | 409.55 | C21H23D8N5O3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY100057S1MG
Encompass_Preferred
Bmy 28674-d8 is the deuterium-labeled isotopologue intended for research use as a stable isotope tracer and internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. Supplied as a 1 mg powdered sample with high purity and defined cold-storage requirements.
  • Deuterium-labeled isotopologue for use as tracer and internal standard.
  • Suitable for quantitative NMR, GC-MS, and LC-MS analysis.
  • High purity (96.13%).
  • Packaged as a 1 mg powder for analytical use.
  • Storage: powder -20°C (stable up to 3 years); in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-15429