Learn More
ATHENA ES BMZERO™, 500 ml
Catalog No. 5000158171
Encompass_Preferred
BRFF-BMZERO™ is a complete serum-free medium comprised of essential amino acids, vitamins, inorganic salts, trace elements and growth factors designed for culturing human breast cell lines. BMZERO™ was developed and has been extensively used for establishing new epithelial cell lines from explants of human breast tissue and to grow immortalized normal breast cell lines. Optimal attached monolayer cell growth can be achieved when used in conjunction with FNC Coating Mix.
Catalog No. 50-001-58171 Supplier ATHENA ES Supplier No. 0401
BRFF-BMZERO™ is a complete serum-free medium comprised of essential amino acids, vitamins, inorganic salts, trace elements and growth factors designed for culturing human breast cell lines. BMZERO™ was developed and has been extensively used for establishing new epithelial cell lines from explants of human breast tissue and to grow immortalized normal breast cell lines. Optimal attached monolayer cell growth can be achieved when used in conjunction with FNC Coating Mix.