Learn More
SAI Infusion Technologies 1cc Syringe with attached 25G0.5" Blunt Needle - Sterile, 100/pack
SDP
Catalog No. NC0878251
Encompass
This sterile 1cc syringe comes with SAI's 25G x 0.5 inch blunt needle already attached, making it easy to maintain sterility while accessing catheters. Ideal for long-term infusion studies requiring flushing and locking of catheters. 100/pk. Product is for laboratory animal research only.
Catalog No. NC0878251 Supplier SAI Infusion Technologies Supplier No. B25501CC100
This sterile 1cc syringe comes with SAI's 25G x 0.5 inch blunt needle already attached, making it easy to maintain sterility while accessing catheters. Ideal for long-term infusion studies requiring flushing and locking of catheters. 100/pk. Product is for laboratory animal research only.