Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BN82002 hydrochloride | 1049740-43-3 | 99.4% | 395.88 | 10 MG
SDP

Catalog No. 5000437095
Encompass_Preferred

BN82002 hydrochloride is a potent, selective, and irreversible inhibitor of the CDC25 phosphatase family. It demonstrates significant inhibitory activity against various CDC25 isoforms including CDC25A, CDC25B2, CDC25B3, CDC25C, and 25C-cat, with IC50 values ranging from 2.4 to 6.3 μM. This compound also exhibits high selectivity, showing approximately 20-fold greater inhibition over CD45 tyrosine phosphatase.

  • Potent and selective CDC25 phosphatase inhibitor
  • Inhibits CDC25A, CDC25B2, CDC25B3, CDC25C, and 25C-cat
  • Demonstrates approximately 20-fold greater selectivity over CD45 tyrosine phosphatase
  • Irreversible inhibitor

Catalog No. 50-004-37095 Supplier Medchemexpress LLC Supplier No. HY112776A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BN82002 hydrochloride is a potent, selective, and irreversible inhibitor of the CDC25 phosphatase family. It demonstrates significant inhibitory activity against various CDC25 isoforms including CDC25A, CDC25B2, CDC25B3, CDC25C, and 25C-cat, with IC50 values ranging from 2.4 to 6.3 μM. This compound also exhibits high selectivity, showing approximately 20-fold greater inhibition over CD45 tyrosine phosphatase.

  • Potent and selective CDC25 phosphatase inhibitor
  • Inhibits CDC25A, CDC25B2, CDC25B3, CDC25C, and 25C-cat
  • Demonstrates approximately 20-fold greater selectivity over CD45 tyrosine phosphatase
  • Irreversible inhibitor

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.