Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bn82002 hydrochloride | 1049740-43-3 | MFCD08276983 | 99.4% | 395.88 | C19H26ClN3O4 | 5 MG
SDP

Catalog No. 5000262305
Encompass_Preferred

BN82002 hydrochloride is an irreversible inhibitor of the CDC25 phosphatase family for laboratory research. It is supplied as the hydrochloride salt and is available as a solid or as a 10 mM solution in DMSO (CAS 1049740-43-3).

  • Potent, selective inhibitor of CDC25 phosphatases.
  • Available as solid and 10 mM solution in DMSO for convenience.
  • High purity: 99.4% (RP-HPLC).
  • Chemical formula C19H26ClN3O4; molecular weight 395.88.
  • Storage: solid at 4°C; in solvent at -80°C (up to 6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.

Catalog No. 50-002-62305 Supplier Medchemexpress LLC Supplier No. HY112776A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BN82002 hydrochloride is an irreversible inhibitor of the CDC25 phosphatase family for laboratory research. It is supplied as the hydrochloride salt and is available as a solid or as a 10 mM solution in DMSO (CAS 1049740-43-3).

  • Potent, selective inhibitor of CDC25 phosphatases.
  • Available as solid and 10 mM solution in DMSO for convenience.
  • High purity: 99.4% (RP-HPLC).
  • Chemical formula C19H26ClN3O4; molecular weight 395.88.
  • Storage: solid at 4°C; in solvent at -80°C (up to 6 months) or -20°C (1 month).
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.