Learn More
SPEX CERTIPREP LLC 3P 8270 Tuning Mix | 1 x 1.5mL
Catalog No. NC1859810
Encompass_Preferred
(NSI by Zeptometrix) 8270 Tuning Mix | 500 ug/mL in Methanol | 1 x 1.5mL
Catalog No. NC1859810 Supplier SPEX CERTIPREP LLC 3P Supplier No. C491
(NSI by Zeptometrix) 8270 Tuning Mix | 500 ug/mL in Methanol | 1 x 1.5mL