Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BNAs in Soil, MilliporeSigma™ Supelco™

Catalog No. 111017291 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
40 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7291 40 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7291 Supplier MilliporeSigma Supelco Supplier No. SQC00340G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

CAS 118-74-1
Type Certified Reference Material
Quantity 40 g
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.