Learn More
Medchemexpress LLC BNC-101 5mg | 5mg
SDP
Catalog No. 5000324985
Encompass_Preferred
BNC-101 is a human-derived antibody expressed in CHO cells that targets GPR49/LGR5 The predicted molecular weight (MW) of BNC-101 is 150 kDa The isotype control for BNC-101 can refer to Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-24985 Supplier Medchemexpress LLC Supplier No. HYP9905605MG
BNC-101 is a human-derived antibody expressed in CHO cells that targets GPR49/LGR5 The predicted molecular weight (MW) of BNC-101 is 150 kDa The isotype control for BNC-101 can refer to Human IgG1 kappa Isotype Control (HY-P99001)