Learn More
World Precision Instrument BNC to Double Banana Connector
SDP
Catalog No. 50822009
Encompass
For wiring any laboratory setup
  • Connector A: Double-banana (male)
  • Connector B: BNC (female)
  • Cable: none
Catalog No. 50-822-009 Supplier World Precision Instrument Supplier No. 13347
For wiring any laboratory setup
  • Connector A: Double-banana (male)
  • Connector B: BNC (female)
  • Cable: none