Learn More
Aviva Systems Biology Corp Bnc2 Antibody - C-terminal region (ARP39297_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39297_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Bnc2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 5031434M05Rik, 8430420F16Rik, MGC124423, MGC124424ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-797