Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bnc375 | 1557240-80-8 | 99.6% | 394.92 g/mol | C19H23ClN2O3S | 5 MG
SDP

Catalog No. 5000262448
Encompass_Preferred

BNC375 is a small-molecule, orally available type I positive allosteric modulator of the α7 nicotinic acetylcholine receptor (α7 nAChR). It demonstrates an EC50 of about 1.9 μM and exhibits CNS drug-like properties, supplied as a high-purity research chemical for in vitro and in vivo pharmacology studies.

  • Potent, selective type I positive allosteric modulator of α7 nAChR.
  • EC50 approximately 1.9 μM in functional assays.
  • High purity suitable for in vitro and in vivo studies.
  • Soluble in DMSO; recommended in vivo formulations provided.
  • Available in milligram-scale quantities for research use.
  • Storage recommendations provided to maintain stability.

Catalog No. 50-002-62448 Supplier Medchemexpress LLC Supplier No. HY1285755MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BNC375 is a small-molecule, orally available type I positive allosteric modulator of the α7 nicotinic acetylcholine receptor (α7 nAChR). It demonstrates an EC50 of about 1.9 μM and exhibits CNS drug-like properties, supplied as a high-purity research chemical for in vitro and in vivo pharmacology studies.

  • Potent, selective type I positive allosteric modulator of α7 nAChR.
  • EC50 approximately 1.9 μM in functional assays.
  • High purity suitable for in vitro and in vivo studies.
  • Soluble in DMSO; recommended in vivo formulations provided.
  • Available in milligram-scale quantities for research use.
  • Storage recommendations provided to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.