Learn More
Medchemexpress LLC BNC375 | 1557240-80-8 | 99.6% | 50 MG
SDP
Catalog No. 5000341887
Encompass_Preferred
BNC375 is a potent, selective, and orally available type I positive allosteric modulator of α7 nAChRs, exhibiting an EC50 of 1.9 μM. It possesses good CNS-agent like properties and has clinical candidate potential. It significantly potentiates the acetylcholine signal without altering rapid receptor desensitization.
  • Potent, selective, and orally available type I positive allosteric modulator of α7 nAChRs.
  • Exhibits good CNS-agent-like properties.
  • Has clinical candidate potential.
  • Significantly potentiates the acetylcholine signal.
  • Achieves full reversal of scopolamine-induced impairment at 1.0 mg/kg in a mouse T-maze model.
  • Minimum effective dose (MED) of 0.03 mg/kg.
  • Plasma half-life (t1/2) of 1.2 hours.
Catalog No. 50-003-41887 Supplier Medchemexpress LLC Supplier No. HY12857550MG
BNC375 is a potent, selective, and orally available type I positive allosteric modulator of α7 nAChRs, exhibiting an EC50 of 1.9 μM. It possesses good CNS-agent like properties and has clinical candidate potential. It significantly potentiates the acetylcholine signal without altering rapid receptor desensitization.
  • Potent, selective, and orally available type I positive allosteric modulator of α7 nAChRs.
  • Exhibits good CNS-agent-like properties.
  • Has clinical candidate potential.
  • Significantly potentiates the acetylcholine signal.
  • Achieves full reversal of scopolamine-induced impairment at 1.0 mg/kg in a mouse T-maze model.
  • Minimum effective dose (MED) of 0.03 mg/kg.
  • Plasma half-life (t1/2) of 1.2 hours.