Learn More
Azzota Corp Benchtop Precision Ph/Mv/C/ F Meter, Ph Range -2.00-16.00Ph, Mv Range: 0 To +/-1999.9Mv, Accuracy: +/- 0.002
Catalog No. 501612909
Encompass_Preferred
Benchtop Precision pH/mV/C/ F Meter, pH Range -2.00-16.00pH, mV Range: 0 to +/-1999.9mV, accuracy: +/- 0.002
Catalog No. 50-161-2909 Supplier Azzota Corp Supplier No. PHMVT921
Benchtop Precision pH/mV/C/ F Meter, pH Range -2.00-16.00pH, mV Range: 0 to +/-1999.9mV, accuracy: +/- 0.002