Learn More
Azzota Corp Benchtop Shaking Incubator, Temp: Amb +5°C To 50°C, Temp. Dis: 0.1°C, Temp. Acc: +/- 0.1°C, Temp. Stab: +/- 0.5°C, Shake S Range: 40 To 300 R/Min, Orbit: 20mm, Time: 1 Min To 99 H 59 M
Catalog No. 501953921
Encompass_Preferred
Benchtop Shaking Incubator, Temp. Range: Ambient +5°C to 50°C, Temp. Display: 0.1°C, Temp. Accuracy: +/- 0.1°C, Temp. Stability: +/- 0.5°C, Shaking Speed Range: 40 to 300 r/min, Orbit: 20mm, Time Range: 1 Min to 99 H 59 M, Shelves: 50mlx42, 100mlx28, 250mlx12, 500mlx11, 1000mlx8, 2000mlx4, Platform Size: 450x450 mm, Machine Size: 590x740x455 mm, Power: 110V/60Hz or 220V 50HZ, Power Consumption: 600W
Catalog No. 50-195-3921 Supplier Azzota Corp Supplier No. BSH4545
Benchtop Shaking Incubator, Temp. Range: Ambient +5°C to 50°C, Temp. Display: 0.1°C, Temp. Accuracy: +/- 0.1°C, Temp. Stability: +/- 0.5°C, Shaking Speed Range: 40 to 300 r/min, Orbit: 20mm, Time Range: 1 Min to 99 H 59 M, Shelves: 50mlx42, 100mlx28, 250mlx12, 500mlx11, 1000mlx8, 2000mlx4, Platform Size: 450x450 mm, Machine Size: 590x740x455 mm, Power: 110V/60Hz or 220V 50HZ, Power Consumption: 600W