Learn More
MSC ALPHATEC SMS DISP CVRALL
Catalog No. 502739499
Encompass
Disposable & Chemical Resistant Coveralls; Garment Style: Coveralls ; Size: 2X-Large ; Material: Microporous Polyethylene Laminate Non-Woven; Polypropylene Non-Woven ; Closure Type: 2-Way Zipper with Storm Flap ; Number Of Pockets: 0.000
Catalog No. 50-273-9499 Supplier MSC Supplier No. 00017640
Disposable & Chemical Resistant Coveralls; Garment Style: Coveralls ; Size: 2X-Large ; Material: Microporous Polyethylene Laminate Non-Woven; Polypropylene Non-Woven ; Closure Type: 2-Way Zipper with Storm Flap ; Number Of Pockets: 0.000