Learn More
MSC ALPHATEC HIVIZ DISP CVRLL
Catalog No. 502739397
Encompass
Disposable & Chemical Resistant Coveralls; Garment Style: Coveralls ; Size: 2X-Large ; Material: Microporous Polyethylene Laminate Non-Woven ; Closure Type: 2-Way Zipper with Storm Flap ; Number Of Pockets: 0.000 ; Cuff Style: Elastic with Thumb-loop
Catalog No. 50-273-9397 Supplier MSC Supplier No. 00288696
Disposable & Chemical Resistant Coveralls; Garment Style: Coveralls ; Size: 2X-Large ; Material: Microporous Polyethylene Laminate Non-Woven ; Closure Type: 2-Way Zipper with Storm Flap ; Number Of Pockets: 0.000 ; Cuff Style: Elastic with Thumb-loop