Learn More
MSC DISP CHEM RESIST CT APRN
Supplier:  MSC 86713377
Encompass
Disposable & Chemical Resistant Aprons; Garment Style: Coat Apron ; Garment Type: Chemical-Resistant ; Garment Length (Inch): 83 ; Garment Width (Inch): 55 ; Material: Microporous Polyethylene Laminate Non-Woven ; Closure Type: Hook & Loop
Catalog No. 50-273-6659