Learn More
Med Vet International Adhesive Strip Nexcare Soft 'N Flex 2 X 4 inch Fabric Rectangle Tan Sterile, 571-08, 8/Box
Catalog No. 502830252
Encompass_Preferred
These 3M™ Nexcare™ ultra-stretch bandages protect minor cuts, blisters and abrasions They are highly flexible to provide coverage on hard to dress areas Latex-free and hypoallergenic, the bandages are gentle Their diamond shape helps build a better seal to keep out pathogens They are breathable to support healing
Catalog No. 50-283-0252 Supplier Med Vet International Supplier No. 1083962BX
These 3M™ Nexcare™ ultra-stretch bandages protect minor cuts, blisters and abrasions They are highly flexible to provide coverage on hard to dress areas Latex-free and hypoallergenic, the bandages are gentle Their diamond shape helps build a better seal to keep out pathogens They are breathable to support healing