Learn More
Med Vet International Dynarex Unna Boot Bandages w/Calamine, 4" X 10 Yards, 12/Case
Catalog No. 502830846
Encompass_Preferred
Unna Boot Bandage uses a non-raveling thread locked gauze, which prevents wound entanglement. For treatment of venous stasis leg ulcers, thrombophlebitis, lymphatic edema, sprains, strains and dislocations Soothing zinc oxide paste formula Plain or with calamine Enhances patient comfort Individually foil wrapped bandages Unique knit construction allows for more uniform distribution of paste Finished edges; will not fray into wound site
Catalog No. 50-283-0846 Supplier Med Vet International Supplier No. 3456
Unna Boot Bandage uses a non-raveling thread locked gauze, which prevents wound entanglement. For treatment of venous stasis leg ulcers, thrombophlebitis, lymphatic edema, sprains, strains and dislocations Soothing zinc oxide paste formula Plain or with calamine Enhances patient comfort Individually foil wrapped bandages Unique knit construction allows for more uniform distribution of paste Finished edges; will not fray into wound site