Learn More
Med Vet International Cohesive Bandage 3M Coban Lf 3 inch X 5 Yard Self-Adherent Closure Tan Sterile Standard Compression, 2083S, 24/Case
Catalog No. 502832225
Encompass_Preferred
The 3M self-adhesive bandage wraps stick to themselves without clips, pins or additional adhesive, offering easy application You can tear the length needed by hand with no need for scissors These 3-inch x 5-yard bandages stay reliably in place to secure primary dressings and splints or to compress and immobilize injury sites The material is porous, lightweight and latex-free for user comfort These Coban bandage wraps come in tan
Catalog No. 50-283-2225 Supplier Med Vet International Supplier No. 388428CS
The 3M self-adhesive bandage wraps stick to themselves without clips, pins or additional adhesive, offering easy application You can tear the length needed by hand with no need for scissors These 3-inch x 5-yard bandages stay reliably in place to secure primary dressings and splints or to compress and immobilize injury sites The material is porous, lightweight and latex-free for user comfort These Coban bandage wraps come in tan