Learn More
Med Vet International Cohesive Bandage 3M Coban Lf 4 inch X 5 Yard Self-Adherent Closure Tan Sterile Standard Compression, 2084S, 18/Case
Catalog No. 502831485
Encompass_Preferred
3M's Coban self-adherent wrap is a multipurpose bandage that secures primary dressings and can provide compression It is self-adherent, ensuring fasteners and clips are unnecessary For convenience, cohesive bandage tears easily without scissors It is lightweight to enhance comfort The breathable material protects the skin integrity Its strong adhesive prevents the tan wrap from shifting to minimize readjustments 4 inches x 5 yards
Catalog No. 50-283-1485 Supplier Med Vet International Supplier No. 386208CS
3M's Coban self-adherent wrap is a multipurpose bandage that secures primary dressings and can provide compression It is self-adherent, ensuring fasteners and clips are unnecessary For convenience, cohesive bandage tears easily without scissors It is lightweight to enhance comfort The breathable material protects the skin integrity Its strong adhesive prevents the tan wrap from shifting to minimize readjustments 4 inches x 5 yards