Learn More
Med Vet International Cohesive Bandage 3M Coban Lf 6 inch X 5 Yard Self-Adherent Closure Tan Sterile Standard Compression, 2086S, 12/Case
Catalog No. 502832201
Encompass_Preferred
Hand tearable – eliminates the need for scissors or cutting Sticks to itself without need for adhesive, pins or clips for fast and easy application Sticks to itself without need for adhesive, pins or clips for fast and easy application Lightweight, porous and comfortable for patients Tan 6 inches x 5 yards
Catalog No. 50-283-2201 Supplier Med Vet International Supplier No. 388429CS
Hand tearable – eliminates the need for scissors or cutting Sticks to itself without need for adhesive, pins or clips for fast and easy application Sticks to itself without need for adhesive, pins or clips for fast and easy application Lightweight, porous and comfortable for patients Tan 6 inches x 5 yards