Learn More
Med Vet International 3M Ace Elastic Bandage, 4 inch X 5 Yard, Clip Detached Closure, Tan, Nonsterile, Standard, Compression, 207433, 10/Box
Catalog No. 502829149
Encompass_Preferred
Innovative clip snaps in place, holds securely and is easily adjusted Provides moderate support to weak, sore muscles and joints Comfortable design is intended for prolonged wear Easy-to-use wrap design Fits discreetly under clothing Wrap conforms to multiple body parts for versatile relief Soft feel is comfortable against your skin Washable and reusable 4 inches x 5 yards
Catalog No. 50-282-9149 Supplier Med Vet International Supplier No. 622BX
Innovative clip snaps in place, holds securely and is easily adjusted Provides moderate support to weak, sore muscles and joints Comfortable design is intended for prolonged wear Easy-to-use wrap design Fits discreetly under clothing Wrap conforms to multiple body parts for versatile relief Soft feel is comfortable against your skin Washable and reusable 4 inches x 5 yards