Learn More
Med Vet International Dynarex High-Strength Pressure Bandage, 4", 100/Case
Catalog No. 502831881
Encompass_Preferred
High Strength Pressure Bandages are ideal for higher compression situations. Combines a thick non-adherent dressing, elastic bandage, pressure applicator and closure clip in one simple unit Quick and easy application Israeli type bandage Single Use Only Not made with natural rubber latex
Catalog No. 50-283-1881 Supplier Med Vet International Supplier No. 3683
High Strength Pressure Bandages are ideal for higher compression situations. Combines a thick non-adherent dressing, elastic bandage, pressure applicator and closure clip in one simple unit Quick and easy application Israeli type bandage Single Use Only Not made with natural rubber latex