Learn More
Med Vet International Convatec Unna Boot Bandage, Without Calamine, 4" X 10 Yards, Each
Catalog No. 502830630
Encompass_Preferred
The kit contains both one Unna-Flex elastic Unna boot and one 4" self-adherent compression bandage. The Unna-Flex Elastic Unna Boot provides the benefit of an Unna Boot without being rigid. The Unna-Flex Elastic Unna Boot is a stretchable, elastic gauze bandage with zinc oxide which is ideal for venous leg ulcer care. The Unna-Flex Elastic Unna Boot is easy to use, flexible and comfortable while conforming to the leg.
Catalog No. 50-283-0630 Supplier Med Vet International Supplier No. SQU650944CSH
The kit contains both one Unna-Flex elastic Unna boot and one 4" self-adherent compression bandage. The Unna-Flex Elastic Unna Boot provides the benefit of an Unna Boot without being rigid. The Unna-Flex Elastic Unna Boot is a stretchable, elastic gauze bandage with zinc oxide which is ideal for venous leg ulcer care. The Unna-Flex Elastic Unna Boot is easy to use, flexible and comfortable while conforming to the leg.