Learn More
Med Vet International Jorvet Esmark Bandage/Tourniquet, 4" x 12', Non-Sterile, J0486AN
Catalog No. 5000495307
Encompass_Preferred
Latex roll that is excellent for bloodless surgical field (i.e., equine lower limb).
Catalog No. 50-004-95307 Supplier Med Vet International Supplier No. JOR486AN
Latex roll that is excellent for bloodless surgical field (i.e., equine lower limb).