Learn More
Med Vet International Medichoice Gauze Roll Bandage, 6-Ply, Sterile, Hypoallergenic, 4.5 inch X 4.1 Yards, White, Gzbn3004, 60/Case
Catalog No. 502830704
Encompass_Preferred
100% cotton gauze, washed and fluff-dried Open, crimped weave for fluid absorption, aeration and comfort Finished edges minimize linting and unraveling Used as a primary absorbent dressing layer or as a secondary dressing to secure and protect primary dressings Ideal for bandaging head, limbs and difficult-to-dress sites MediChoice Rolled Gauze Bandages are comprised of 100% cotton gauze, and are ideal for bandaging head, limbs and difficult-to dress sites. Each of these 6-ply bandages measure 4.5 inch X 4.1 Yards and are sterile. MediChoice is Owens & Minor's brand of medical-surgical products and durable medical equipment that is expanding to meet the changing needs of healthcare. MediChoice, your choice for quality, value and unmatched service
Catalog No. 50-283-0704 Supplier Med Vet International Supplier No. GZBN3004
100% cotton gauze, washed and fluff-dried Open, crimped weave for fluid absorption, aeration and comfort Finished edges minimize linting and unraveling Used as a primary absorbent dressing layer or as a secondary dressing to secure and protect primary dressings Ideal for bandaging head, limbs and difficult-to-dress sites MediChoice Rolled Gauze Bandages are comprised of 100% cotton gauze, and are ideal for bandaging head, limbs and difficult-to dress sites. Each of these 6-ply bandages measure 4.5 inch X 4.1 Yards and are sterile. MediChoice is Owens & Minor's brand of medical-surgical products and durable medical equipment that is expanding to meet the changing needs of healthcare. MediChoice, your choice for quality, value and unmatched service