Learn More
Cayman Chemical BngtrIfluoroacetate salt 10mg
SDP
Catalog No. 502915068
Encompass_Preferred
An antimicrobial peptide; active against various Gram-negative and Gram-positive bacteria, including MRSA (MICs = 4-64 UG/ml); synergistic with ampicillin, amoxicillin, and novobiocin against P. aeruginosa; suppresses the development of resistance to kanamycin and ampicillin in E. coli; increases the survival rate of E. tarda-infected O. laptipes
Catalog No. 50-291-5068 Supplier Cayman Chemical Supplier No. 3695910 MG
An antimicrobial peptide; active against various Gram-negative and Gram-positive bacteria, including MRSA (MICs = 4-64 UG/ml); synergistic with ampicillin, amoxicillin, and novobiocin against P. aeruginosa; suppresses the development of resistance to kanamycin and ampicillin in E. coli; increases the survival rate of E. tarda-infected O. laptipes