Learn More
Aviva Systems Biology Corp BNIP1 Antibody - C-terminal region (ARP59521_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59521_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BNIP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human BNIP1. Alternate names: NIP1, SEC20, TRG-8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-724