Learn More
Aviva Systems Biology Corp BNIPL Antibody - middle region (ARP52577_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52577_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BNIPL. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human BNIPL. Alternate names: BNIP-S, BNIPL-1, BNIPL-2, PP753ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-734