Learn More
Bachem BNP-45 (mouse) Trifluoroacetate
SDP
Supplier:  Bachem 4095506.0500
Encompass_Preferred
BNP-45 (mouse) Trifluoroacetate, H-Ser-Gln-Gly-Ser-Thr-Leu-Arg-Val-Gln-Gln-Arg-Pro-Gln-Asn-Ser-Lys-Val-Thr-His-Ile-Ser-Ser-Cys-Phe-Gly-His-Lys-Ile-Asp-Arg-Ile-Gly-Ser-Val-Ser-Arg-Leu-Gly-Cys-Asn-Ala-Leu-Lys-Leu-Leu-OH, 1816939-52-2, C209H354N70O63S2, Mr 4919.61, 0.5mg. Brain Natriuretic Peptide-45 (mouse), BNP (1-45), mouse.
Catalog No. 50-194-6768