Learn More
Enzo Life Sciences Benzylphosphonic acid-(AM)2 (10mg). CAS: 200716-68-3
SDP
Catalog No. 502010631
Encompass_Preferred
Benzylphosphonic acid-(AM)2 is a bis-acetoxymethyl ester (AM) derivative of benzylphosphonic acid which is cell permeable and is converted to the active phosphonic acid via intracellular esterase-catalyzed cleavage of the AM esters. Purity: ≥98% (TLC). Solubility: Soluble in ethanol (20 mg/ml) or DMSO (20 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0631 Supplier Enzo Life Sciences Supplier No. BMLEI2450010
Add to Cart
Add to Cart
Benzylphosphonic acid-(AM)2 is a bis-acetoxymethyl ester (AM) derivative of benzylphosphonic acid which is cell permeable and is converted to the active phosphonic acid via intracellular esterase-catalyzed cleavage of the AM esters. Purity: ≥98% (TLC). Solubility: Soluble in ethanol (20 mg/ml) or DMSO (20 mg/ml). Long Term Storage: -20°C.