Learn More
CPC Scientific H-Ser-Gln-Gly-Ser-Thr-Leu-Arg-Val-Gln-Gln-Arg-Pro-Gln-Asn-Ser-Lys-Val-Thr-His-Ile-Ser-Ser-Cys-Phe-Gly-His-Lys-Ile-Asp-Arg-Ile-Gly-Ser-Val-Ser-Arg-Leu-Gly-Cys-Asn-Ala-Leu-Lys-Leu-Leu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific NATR018B
Encompass
ONE-LETTER SEQUENCE: SQGSTLRVQQRPQNSKVTHISSCFGHKIDRIGSVSRLGCNALKLL (Cys23 and 39 bridge)MOLECULAR FORMULA: C209H354N70O63S2MOLECULAR WEIGHT:4919.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [123337-89-3], SYNONYMS: BNP-45 (mouse), Brain Natriuretic Peptide-45 (mouse)RESEARCH AREA: CardiovascularREFERENCES: M. Aburaya et al., BBRC, 163, 226 (1989)
Catalog No. 50-210-9875